Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLI, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115570 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-570 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-570 Supplier Abnova Corporation Supplier No. PAB12066
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of POLI.

Specifications

Antigen POLI
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of POLI.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In antiserum (0.02% sodium azide)
Gene POLI
Gene Alias RAD30B/RAD3OB
Gene Symbols POLI
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human POLI.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11201
Test Specificity This antibody is specific to human DNA Polymerase Iota.
Target Species Human, Mouse
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.