Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Por, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115481 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-481 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-481 Supplier Abnova Corporation Supplier No. PAB11972

Rabbit polyclonal antibody raised against native Por.

Specifications

Antigen Por
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native Por.
Dilution Western Blot (1:1000) Immunocytochemistry (1:500-1:1000) Immunoprecipitation (1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide, 50% glycerol)
Gene Por
Gene Symbols Por
Host Species Rabbit
Immunogen Native purified Por from rat liver.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29441
Test Specificity This antibody detects a 78 KDa protein on SDS-PAGE immunoblots, corresponding to the molecular weight of cytochrome P450 reductase.
Target Species Avian, Bovine, Canine, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rat, Sheep
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.