Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POSTN, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant POSTN.
Supplier: Abnova Corporation PAB13770
Specifications
POSTN | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant POSTN. | |
Lyophilized from 50mM phosphate buffer, 100mM NaCl, pH 7.2 | |
MGC119510/MGC119511/OSF-2/PDLPOSTN/PN/RP11-412K4.1/periostin | |
Rabbit | |
50 μg | |
Primary | |
Recognizes human, rat and chicken OSF-2. | |
Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Western Blot (0.1-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:5000) The optimal working dilution should be determined by the end user. | |
POSTN | |
POSTN | |
Recombinant protein corresponding to human POSTN. | |
RUO | |
10631 | |
Chicken, Human, Rat | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction