Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Postn Rabbit anti-Chicken, Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89118746 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-746 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-746 Supplier Abnova Corporation Supplier No. PAB16942

Rabbit polyclonal antibody raised against synthetic peptide of Postn.

Specifications

Antigen Postn
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Postn.
Dilution Western Blot (1:1000) Immunohistochemistry (1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene Postn
Gene Alias AI747096/OSF-2/Osf2/PN/Periostin/peri
Gene Symbols Postn
Host Species Rabbit
Immunogen A synthetic peptide corresponding to fasciclin domain 1 of mouse Postn.
Purification Method Affinity purification
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50706
Test Specificity This antibody has been directly tested for reactivity in chicken, human, mouse and rat. Based on the homology of the immunogen with other species, we expect that this antibody will recognize periostin in most, if not all, species of mammals, birds, amphibians, and fishes. specific to three periostin bands around ∼93 KDa in mouse lung extract. The immunogenic peptide is present in all known splice variants of periostin and therefore this antibody recognizes all known molecular forms of periostin. This antibody also works well for immunohistochemistry on paraformaldehyde-fixed sections with a simple antigenretrieval protocol (incubate slides for 20 minutes at 90 °C in 10 mM sodium citrate (pH 6.0)/ 0.1% Tween-20).
Target Species Chicken, Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.