Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POU2AF1, Rabbit, Clone: SP92, Abnova™

Catalog No. 89108359 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-359 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-359 Supplier Abnova Corporation Supplier No. MAB10044

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit monoclonal antibody raised against synthetic peptide of POU2AF1.

Specifications

Antigen POU2AF1
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone SP92
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of POU2AF1.
Dilution Western Blot (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.6 (1% BSA, 0.09% sodium azide)
Gene POU2AF1
Gene Alias BOB1/OBF-1/OBF1/OCAB
Gene Symbols POU2AF1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human POU2AF1.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5450
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.