Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POU3F2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89127299 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-299 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-299 Supplier Abnova Corporation Supplier No. PAB25679
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of POU3F2.

POU3F2 belongs to a large family of transcription factors that bind to the octameric DNA sequence ATGCAAAT. Most of these proteins share a highly homologous region, referred to as the POU domain, that occurs in several mammalian transcription factors, including the octamer-binding proteins Oct1 (POU2F1; MIM 164175) and Oct2 (POU2F2; MIM 164176) and the pituitary protein Pit1 (PIT1; MIM 173110). Class III POU genes are expressed predominantly in the central nervous system (CNS). It is likely that CNS-specific transcription factors such as these play an important role in mammalian neurogenesis by regulating their diverse patterns of gene expression (Schreiber et al., 1993 [PubMed 8441633]; Atanasoski et al., 1995 [PubMed 7601453]).[supplied by OMIM

Specifications

Antigen POU3F2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of POU3F2.
Dilution Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene POU3F2
Gene Alias BRN2/OCT7/OTF7/POUF3
Gene Symbols POU3F2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 18 amino acids at C-terminus of human POU3F2.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5454
Test Specificity At least two isoforms are known to exist. This antibody will recognize both isoforms. POU3F2 antibody is predicted to not cross-react with other members of the POU domain class 3 family.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.