Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPAPDC1A, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120681 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-681 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-681 Supplier Abnova Corporation Supplier No. PAB19155
Add to Cart
Add to Cart

Rabbit polyclonal antibody rasied against synthetic peptide of PPAPDC1A.

Specifications

Antigen PPAPDC1A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody rasied against synthetic peptide of PPAPDC1A.
Dilution Western blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene PPAPDC1A
Gene Accession No. NP_001025230
Gene Alias DPPL2/MGC120299/MGC120300/PPAPDC1
Gene Symbols PPAPDC1A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 14 amino acids near C-terminus of human PPAPDC1A.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196051
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.