Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPARG Rabbit anti-Dog, Guinea pig, Hamster, Human, Macaque, Mink, Mouse, Orangutan, Pig, Rabbit, Rat, Squirrel, Polyclonal Antibody, Abnova™

Catalog No. 89114956 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-956 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-956 Supplier Abnova Corporation Supplier No. PAB11247
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of PPARG.

This gene encodes a member of the peroxisome proliferator-activated receptor (PPAR) subfamily of nuclear receptors. PPARs form heterodimers with retinoid X receptors (RXRs) and these heterodimers regulate transcription of various genes. Three subtypes of PPARs are known: PPAR-alpha, PPAR-delta, and PPAR-gamma. The protein encoded by this gene is PPAR-gamma and is a regulator of adipocyte differentiation. Additionally, PPAR-gamma has been implicated in the pathology of numerous diseases including obesity, diabetes, atherosclerosis and cancer. Alternatively spliced transcript variants that encode different isoforms have been described. [provided by RefSeq

Specifications

Antigen PPARG
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of PPARG.
Dilution ELISA (1:8000-1:32000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene PPARG
Gene Alias CIMT1/NR1C3/PPARG1/PPARG2/PPARgamma
Gene Symbols PPARG
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 255 -268 of human PPARG.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5468
Test Specificity This affinity purified antibody is directed against human PPAR gamma protein. Expect reactivity against human (gamma1 and gamma2), mouse (gamma1 and gamma2), rat, squirrel, orangutan, duck (gamma1), macaque (gamma1-gamma7), boar (gamma1a-1d), mink, guinea pig, rabbit (gamma1 and gamma3), dog and hamster sources based on 100% homology for the immunogen sequence. No reactivity is expected against other subtypes of PPAR.
Target Species Canine, Guinea Pig, Hamster, Human, Macaque, Mink, Mouse, Orangutan, Pig, Rabbit, Rat, Squirrel
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.