Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPM1A/PPM1B, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89137647 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-647 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-647 Supplier Abnova Corporation Supplier No. PAB13655
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of PPM1A/PPM1B.

Sequence: RYGLSSMGWRVE

Specifications

Antigen PPM1A/PPM1B
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of PPM1A/PPM1B.
Dilution Western Blot (5-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to rabbit PPM1A/PPM1B.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This is a rabbit polyclonal antibody is specific to serine/threonine proten phosphatase 2C alpha/beta(PP 2C alpha /beta) corresponding to a specific peptide immunogen to enhance reactivity and minimize cross reactivity. This antibody does not cross-react with other protein phosphatase isoforms.
Target Species Bovine, Human, Mouse, Rabbit, Rat
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.