Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1CB Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114839 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-839 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-839 Supplier Abnova Corporation Supplier No. PAB10412
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of PPP1CB.

The protein encoded by this gene is one of the three catalytic subunits of protein phosphatase 1 (PP1). PP1 is a serine/threonine specific protein phosphatase known to be involved in the regulation of a variety of cellular processes, such as cell division, glycogen metabolism, muscle contractility, protein synthesis, and HIV-1 viral transcription. Mouse studies suggest that PP1 functions as a suppressor of learning and memory. Two alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq

Sequence: PRTANPPKKR

Specifications

Antigen PPP1CB
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of PPP1CB.
Dilution Western blot (5 to 10 ug/mL) Immunoprecipitation (10 to 15 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Gene PPP1CB
Gene Alias MGC3672/PP-1B/PPP1CD
Gene Symbols PPP1CB
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH ) corresponding to amino acids 318-327 of PPP1CB.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5500
Target Species Bovine, Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.