Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1R1B Rabbit anti-Bovine, Chicken, Clawed frog, Dog, Human, Mouse, Rat, Zebra fish, Polyclonal Antibody, Abnova™

Catalog No. 89134282 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-282 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-282 Supplier Abnova Corporation Supplier No. PAB9626
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Ppp1r1b.

Specifications

Antigen PPP1R1B
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Ppp1r1b.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA)
Gene Ppp1r1b
Gene Alias Darpp-32/Darpp32
Gene Symbols Ppp1r1b
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to N-terminus rat Ppp1r1b.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360616
Target Species Bovine, Canine, Chicken, Frog, Human, Mouse, Rat, Zebrafish
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.