Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1R1B Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89120029 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-029 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-029 Supplier Abnova Corporation Supplier No. PAB18466
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of PPP1R1B.

Midbrain dopaminergic neurons play a critical role in multiple brain functions, and abnormal signaling through dopaminergic pathways has been implicated in several major neurologic and psychiatric disorders. One well-studied target for the actions of dopamine is DARPP32. In the densely dopamine- and glutamate-innervated rat caudate-putamen, DARPP32 is expressed in medium-sized spiny neurons (Ouimet and Greengard, 1990 [PubMed 2191086]) that also express dopamine D1 receptors (Walaas and Greengard, 1984 [PubMed 6319627]). The function of DARPP32 seems to be regulated by receptor stimulation. Both dopaminergic and glutamatergic (NMDA) receptor stimulation regulate the extent of DARPP32 phosphorylation, but in opposite directions (Halpain et al., 1990 [PubMed 2153935]). Dopamine D1 receptor stimulation enhances cAMP formation, resulting in the phosphorylation of DARPP32 (Walaas and Greengard, 1984 [PubMed 6319627]); phosphorylated DARPP32 is a potent protein phosphatase-1 (see MIM 176875) inhibitor (Hemmings et al., 1984 [PubMed 6087160]). NMDA receptor stimulation elevates intracellular calcium, which leads to activation of calcineurin and dephosphorylation of phospho-DARPP32, thereby reducing the phosphatase-1 inhibitory activity of DARPP32 (Halpain et al., 1990 [PubMed 2153935]).[supplied by OMIM

Specifications

Antigen PPP1R1B
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of PPP1R1B.
Dilution Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:20000) The optimal working dilution should be determined by the end user.
Formulation In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
Gene PPP1R1B
Gene Alias DARPP-32/DARPP32/FLJ20940
Gene Symbols PPP1R1B
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues surrounding T34 of human PPP1R1B.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84152
Test Specificity This antibody is specific to ATF2.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.