Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1R3D, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89125463 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-125-463 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-125-463 Supplier Abnova Corporation Supplier No. PAB23755
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant PPP1R3D.

Phosphorylation of serine and threonine residues in proteins is a crucial step in the regulation of many cellular functions ranging from hormonal regulation to cell division and even short-term memory. The level of phosphorylation is controlled by the opposing actions of protein kinases and protein phosphatases. Protein phosphatase 1 (PP1) is 1 of 4 major serine/threonine-specific protein phosphatases which have been identified in eukaryotic cells. PP1 associates with various regulatory subunits that dictate its subcellular localization and modulate its substrate specificity. Several subunits that target PP1 to glycogen have been identified. This gene encodes a glycogen-targeting subunit of PP1. [provided by RefSeq

Sequence: ELAQVKVFNAGDDPSVPLHVLSRLAINSDLCCSSQDLEFTLHCLVPDFPPPVEAADFGERLQRQLVCLERVTCSDL

Specifications

Antigen PPP1R3D
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant PPP1R3D.
Dilution Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene PPP1R3D
Gene Alias DKFZp781L2441/PPP1R6
Gene Symbols PPP1R3D
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human PPP1R3D.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5509
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.