Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP2R1A Rabbit anti-Bovine, Human, Mouse, Rabbit, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89137648 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-648 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-648 Supplier Abnova Corporation Supplier No. PAB13656
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of PPP2R1A.

Sequence: DDSLYPIAVLIDGlu

Specifications

Antigen PPP2R1A
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of PPP2R1A.
Dilution Western Blot (1:500-1:1000) Immunocytochemistry (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 7-19 of rabbit PPP2R1A.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This is a characterized rabbit anti-peptide antibodies corresponding to a selected region of a specific mammalian protein serine/threonine phosphatase. These antibodies are made against specific peptides to enhance reactivity and minimize cross reactivity. Reacts with human, mouse, rat and bovine PP 2A/A (65 KDa).
Target Species Bovine, Human, Mouse, Rabbit, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.