Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP2R2A Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114843 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-843 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-843 Supplier Abnova Corporation Supplier No. PAB10418
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of PPP2R2A.

The product of this gene belongs to the phosphatase 2 regulatory subunit B family. Protein phosphatase 2 is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes an alpha isoform of the regulatory subunit B55 subfamily. [provided by RefSeq

Sequence: FSQVLGAVDDDVAE

Specifications

Antigen PPP2R2A
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of PPP2R2A.
Dilution Western blot (5 to 10 ug/mL) Immunocy to chemistry (5 to 10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Gene PPP2R2A
Gene Alias B55-ALPHA/B55A/FLJ26613/MGC52248/PR52A/PR55A
Gene Symbols PPP2R2A
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH ) corresponding to amino acids 14-27 of PPP2R2A.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5520
Target Species Bovine, Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.