Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRAGMIN, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115072 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-072 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-072 Supplier Abnova Corporation Supplier No. PAB1143

Rabbit polyclonal antibody raised against synthetic peptide of PRAGMIN.

Sequence: KFAEKAVDRRRGVELED

Specifications

Antigen PRAGMIN
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description homolog of rat pragma of Rnd2
Formulation In rabbit sera (50% glycerol, 0.025% sodium azide)
Gene PRAGMIN
Gene Alias DKFZp761P0423
Gene Symbols PRAGMIN
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 1361-1377 of human PRAGMIN.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157285
Target Species Human, Mouse
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.