Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRC1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89127112 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-112 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-112 Supplier Abnova Corporation Supplier No. PAB25495
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of PRC1.

This gene encodes a protein that is involved in cytokinesis. The encoded protein is at high level during S and G2/M and drop dramatically after cell exit mitosis and enter G1. It is located in the nucleus during interphase, and becomes associated with mitotic spindles in a highly dynamic manner during mitosis, and localizes to the cell mid-body during cytokinesis. This protein has been shown to be a substrate of several cyclin-dependent kinases (CDKs). At least three alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq

Specifications

Antigen PRC1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of PRC1.
Dilution Western Blot (0.5-1 ug/mL) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene PRC1
Gene Alias ASE1/MGC1671/MGC3669
Gene Symbols PRC1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal region of human PRC1.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9055
Test Specificity At least three alternatively spliced transcript variants encoding distinct isoforms have been observed.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.