Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRDM2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89125538 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-125-538 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-125-538 Supplier Abnova Corporation Supplier No. PAB2383
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant Prdm2.

Specifications

Antigen PRDM2
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Prdm2.
Dilution Immunohistochemistry (1:100-500) Immunoprecipitation The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Prdm2
Gene Accession No. NM_012231;NM_015866
Gene Alias Riz
Gene Symbols Prdm2
Host Species Rabbit
Immunogen Recombinant GST fusion protein corresponding to amino acids 245-573 of rat Prdm2.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 313678
Test Specificity This antibody also recognizes the isoform beta (Riz2) of PRDM2 (185 KDa).
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.