Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRDM5, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114167 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-167 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-167 Supplier Abnova Corporation Supplier No. PAB0353
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of PRDM5.

The protein encoded by this gene is a transcription factor of the PR-domain protein family. It contains a PR-domain and multiple zinc finger motifs. Transcription factors of the PR-domain family are known to be involved in cell differentiation and tumorigenesis. [provided by RefSeq

Specifications

Antigen PRDM5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of PRDM5.
Dilution The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.05% sodium azide, 0.2% gelatin
Gene PRDM5
Gene Alias PFM2
Gene Symbols PRDM5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 100-200 of human PRDM5.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11107
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.