Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRDX2 Rabbit anti-Hamster, Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89116839 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-839 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-839 Supplier Abnova Corporation Supplier No. PAB13782

Rabbit polyclonal antibody raised against recombinant Prdx2.

Specifications

Antigen PRDX2
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant Prdx2.
Dilution Western Blot (1:500-1:2000) Immunohistochemistry (Frozen sections) (1:100-1:500) The optimal working dilution should be determined by the end user.
Formulation In antiserum (0.05% sodium azide)
Gene Prdx2
Gene Alias AL022839/Band-8/NkefB/PRP/PrxII/TDX1/TPx/TPx-B/TR/TSA/Tdpx1/Torin
Gene Symbols Prdx2
Host Species Rabbit
Immunogen Recombinant protein corresponding to mouse Prdx2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21672
Test Specificity Recognizes Peroxiredoxin II (PrxII). Does NOT cross react with PrxI.
Target Species Hamster, Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.