Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRDX3, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115509 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-509 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-509 Supplier Abnova Corporation Supplier No. PAB1200

Rabbit polyclonal antibody raised against partial recombinant PRDX3.

This gene encodes a protein with antioxidant function and is localized in the mitochondrion. This gene shows significant nucleotide sequence similarity to the gene coding for the C22 subunit of Salmonella typhimurium alkylhydroperoxide reductase. Expression of this gene product in E. coli deficient in the C22-subunit gene rescued resistance of the bacteria to alkylhydroperoxide. The human and mouse genes are highly conserved, and they map to the regions syntenic between mouse and human chromosomes. Sequence comparisons with recently cloned mammalian homologues suggest that these genes consist of a family that is responsible for regulation of cellular proliferation, differentiation, and antioxidant functions. Two transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen PRDX3
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant PRDX3.
Dilution Western Blot (1:1800) Immunoprecipitation (1 ul) The optimal working dilution should be determined by the end user.
Formulation In HEPES, 150mM NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide)
Gene PRDX3
Gene Alias AOP-1/AOP1/MER5/MGC104387/MGC24293/PRO1748/SP-22
Gene Symbols PRDX3
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 63-256 of human PRDX3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10935
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.