Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prei4 (Gpcpd1), Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117320 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-320 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-320 Supplier Abnova Corporation Supplier No. PAB14875
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Gpcpd1.

Specifications

Antigen Prei4 (Gpcpd1)
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Gpcpd1.
Dilution Immunohistochemistry (5-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.1% proclin, 2% Block Ace)
Gene Prei4
Gene Alias 2310004G06Rik/2310032D16Rik/AU015213/mKIAA1434
Gene Symbols Prei4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to mouse Gpcpd1.
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 74182
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.