Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRICKLE4, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89127371 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-371 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-371 Supplier Abnova Corporation Supplier No. PAB25747
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of PRICKLE4.

C6ORF49 is a member of the LIM domain protein family (Teufel et al., 2005 [PubMed 15702247]).[supplied by OMIM

Specifications

Antigen PRICKLE4
Applications ELISA, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of PRICKLE4.
Dilution Western Blot (0.25 ug/mL) Immunocytochemistry (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene PRICKLE4
Gene Alias C6orf49/MGC120398/OBTP/OEBT
Gene Symbols PRICKLE4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 18 amino acids at N-terminus of human PRICKLE4.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29964
Test Specificity Two isoforms of PRICKLE4 are known to exist. PRICKLE4 antibody is predicted to not cross-react with other PRICKLE protein family members.
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.