Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKACA/PRKACB, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89130701 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-701 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-701 Supplier Abnova Corporation Supplier No. PAB29264
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of human PRKACA/PRKACB.

Specifications

Antigen PRKACA/PRKACB
Applications Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of human PRKACA/PRKACB.
Dilution Immunofluorescence (1:100∼1:200) Immunohistochemistry (1:50∼1:100) Western Blot (1:500∼1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
Gene Alias MGC102831/MGC48865/PKACA
Gene Symbols PRKACA
Host Species Rabbit
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T197 of human PRKACA/PRKACB.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5566/5567
Test Specificity PRKACA/PRKACB (phospho T197) polyclonal antibody detects endogenous levels of human PRKACA/PRKACB only when phosphorylated at threonine 197.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.