Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKCE Rabbit anti-Rat, Polyclonal Antibody, Abnova™

Catalog No. 89116780 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-780 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-780 Supplier Abnova Corporation Supplier No. PAB13643
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Prkce.

Sequence: KGFSYFGEDLMP

Specifications

Antigen PRKCE
Applications ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Prkce.
Dilution Western Blot (1:250) Immunocytochemistry (1:250) Immunohistochemistry (1:250) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.05% sodium azide
Gene Prkce
Gene Alias Pkce
Gene Symbols Prkce
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 726-737 of rat Prkce.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29340
Test Specificity Based on known homologies is expected to react with the PKC ortholog in most mammalian species. These rabbit polyclonal antibodies are made against specific peptide regions of the PKC epsilon protein. By using specific peptides for the PKC delta that has effectively minimized cross reactivity between the various isoforms. These antibodies are useful in the screening of PKC epsilon proteins in various biological tissues and tumorigenesis studies.
Target Species Rat
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.