Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKCH Rabbit anti-Rat, Polyclonal Antibody, Abnova™

Catalog No. 89116782 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-782 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-782 Supplier Abnova Corporation Supplier No. PAB13647
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Prkch.

Specifications

Antigen PRKCH
Applications ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Prkch.
Dilution Western Blot (1:250) Immunocytochemistry (1:250) Immunohistochemistry (1:250) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.05% sodium azide
Gene Prkch
Gene Alias MGC93383
Gene Symbols Prkch
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of rat Prkch.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81749
Test Specificity Based on known homologies is expected to react with the PKC ortholog in most mammalian species. These rabbit polyclonal antibodies are made against specific peptide regions of the PKC eta protein. By using specific peptides for the PKC eta that has effectively minimized cross reactivity between the various isoforms.
Target Species Rat
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.