Learn More
PRKD1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PRKD1.
Supplier: Abnova Corporation PAB26697
Description
Members of the protein kinase C (PKC) family function in many extracellular receptor-mediated signal transduction pathways. See PRKCA (MIM 176960) for further background information. The PRKCM gene encodes a cytosolic serine-threonine kinase that binds to the trans-Golgi network and regulates the fission of transport carriers specifically destined to the cell surface.[supplied by OMIM
Sequence: E-K-Sp-F-RSpecifications
PRKD1 | |
Polyclonal | |
Unconjugated | |
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
PRKD1 | |
PRKD1 | |
A synthetic peptide corresponding to residues surrounding S738 of human PRKD1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of PRKD1. | |
In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
PKC-MU/PKCM/PKD/PRKCM | |
Rabbit | |
Affinity chromatography | |
RUO | |
5587 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only