Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKDC (phospho T2609), Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114788 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-788 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-788 Supplier Abnova Corporation Supplier No. PAB10324
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of PRKDC.

The PRKDC gene encodes the catalytic subunit of a nuclear DNA-dependent serine/threonine protein kinase (DNA-PK). The second component is the autoimmune antigen Ku (MIM 152690), which is encoded by the G22P1 gene on chromosome 22q. On its own, the catalytic subunit of DNA-PK is inactive and relies on the G22P1 component to direct it to the DNA and trigger its kinase activity; PRKDC must be bound to DNA to express its catalytic properties.[supplied by OMIM

Specifications

Antigen PRKDC
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of PRKDC.
Dilution ELISA (1:300000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene PRKDC
Gene Alias DNA-PKcs/DNAPK/DNPK1/HYRC/HYRC1/XRCC7/p350
Gene Symbols PRKDC
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding T2609 of human PRKDC.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5591
Test Specificity This phospho-specific polyclonal antibody reacts with phosphorylated pT2609 of human DNA PKcs. Reactivity with non-phosphorylated human DNA PKcs is minimal.
Target Species Canine, Chicken, Chimpanzee, Equine, Human, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.