Learn More
PRLHR Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PRLHR.
Supplier: Abnova Corporation PAB16337
Description
PRLHR is a 7-transmembrane domain receptor for prolactin-releasing hormone (PRLH; MIM 602663) that is highly expressed in anterior pituitary (Ozawa et al., 2002 [PubMed 11923475]).[supplied by OMIM
Specifications
PRLHR | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of PRLHR. | |
In PBS (0.09% sodium azide) | |
GPR10/GR3/MGC126539/MGC126541/PrRPR | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
2834 | |
Human, Mouse, Rat | |
Liquid |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) The optimal working dilution should be determined by the end user. | |
PRLHR | |
PRLHR | |
A synthetic peptide (conjugated with KLH) corresponding to human PRLHR. | |
50 μg | |
Primary | |
2nd extracellular domain of human. | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only