Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-Pro-Biglycan, Polyclonal,

Catalog No. ABT270MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABT270MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABT270MI Supplier MilliporeSigma™ Supplier No. ABT270
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Pro-Biglycan in Human, Mouse samples, and it is validated for Immunohistochemistry, Western Blotting

Biglycan (UniProt P21810; also known as Biglycan proteoglycan, Bone/cartilage proteoglycan I, Dermatan sulphate proteoglycan I, PG-S1, Small leucine-rich protein 1A) is encoded by the BGN (also known as DSPG1, PGI, SLRR1A) gene (Entrez Gene ID 633) in human. Biglycan is a pericellular matrix proteoglycan structurally related to two other small proteoglycans, decorin and fibromodulin. Biglycan and decorin are thought to be the result of a gene duplication. Decorin contains one attached glycosaminoglycan chain, while biglycan contains two chains, and therefore the name biglycan. Biglycan is involved in collagen fibrils assembly and muscle regeneration. Biglycan is essential for the assembly of dystrophin-associated protein complex, and interacts with several proteins linked to muscular dystrophy, including alpha-dystroglycan, alpha- and gamma-sarcoglycan and collagen VI. Biglycan is initially translated as a preproprotein with an N-terminal signal peptide (aa1-19) and propeptide (aa20-37) sequence. Sequential removal of the signal peptide and the propeptide sequence produces probiglycan and mature biglycan, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pro-Biglycan
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Formulation Rabbit polyclonal antibody serum with 0.05% sodium azide.
Gene Accession No. P21810
Gene Symbols BGN; SLRR1A
Host Species Rabbit
Immunogen LPH-conjugated linear peptide corresponding to the signal and propeptide of human Pro-Biglycan.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001702
Target Species Human, Mouse
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.