Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-Proline-rich Protein 14, Polyclonal,

Catalog No. ABE1398MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1398MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1398MI Supplier MilliporeSigma™ Supplier No. ABE1398
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Proline-rich Protein 14 in Human, Mouse, Rat samples, and it is validated for Western Blotting

Proline-rich protein 14 (UniProt Q9BWN1) is encoded by the PRR14 gene in human (Gene ID 78994). Proline-rich protein 14 participates in the tethering of heterochromatin to the inner nuclear periphery by associating with the nuclear lamina and attaching to heterochromatin through its binding partner, heterochromatin protein 1 (HP1). After disassembly early in mitosis, Proline-rich protein 14 reassembles via binding to anaphase chromosomes through HP1, then via association with the nuclear lamina in telophase. PRR14 may thereby play a role in specifying HP1-bound heterochromatin for reattachment to the nuclear lamina at mitotic exit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Proline-rich Protein 14
Applications Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q9BWN1
Gene Symbols PRR14
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human Proline-rich Protein 14.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_076936
Target Species Human, Mouse, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.