Learn More
PROS1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native PROS1.
Supplier: Abnova Corporation PAB19905
Description
This gene encodes a vitamin K-dependent plasma protein that functions as a cofactor for the anticoagulant protease, activated protein C (APC) to inhibit blood coagulation. It is found in plasma in both a free, functionally active form and also in an inactive form complexed with C4b-binding protein. Mutations in this gene result in autosomal dominant hereditary thrombophilia. An inactive pseudogene of this locus is located at an adjacent region on chromosome 3. [provided by RefSeq
Specifications
| PROS1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against native PROS1. | |
| In PBS, pH 7.5 (0.02% sodium azide, 50% glycerol (v/v)) | |
| PROS/PS21/PS22/PS23/PS24/PS25/PSA | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 5627 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| PROS1 | |
| PROS1 | |
| Native purified PROS1 from human plasma. | |
| 150 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only