Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein Kinase Consensus Sequence, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89137643 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-643 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-643 Supplier Abnova Corporation Supplier No. PAB13649
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Protein Kinase Consensus Sequence.

Sequence: GTPEYLAPE

Specifications

Antigen Protein Kinase Consensus Sequence
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Protein Kinase Consensus Sequence.
Dilution Western Blot (1:50) The optimal working dilution should be determined by the end user.
Formulation In PBS
Host Species Rabbit
Immunogen A synthetic peptide corresponding to Protein Kinase Consensus Sequence.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity It is excellent for Western blot detection of protein kinases containing this sequence.
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.