Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRPH Rabbit anti-Human, Mammals, Polyclonal Antibody, Abnova™

Catalog No. 89115612 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-612 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-612 Supplier Abnova Corporation Supplier No. PAB12115
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against full-length recombinant Prph.

Specifications

Antigen PRPH
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant Prph.
Dilution ELISA (1:1000) Immunocytochemistry (1:1000) Immunofluorescence (1:1000) Western Blot (1:5000) The optimal working dilution should be determined by the end user.
Formulation In antiserum
Gene Prph
Gene Alias Prph1
Gene Symbols Prph
Host Species Rabbit
Immunogen Recombinant protein corresponding to full length rat Prph.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19132
Test Specificity Specifically recognizes peripherin, showing no cross reactivity with other Class III intermediate filament proteins.
Target Species Human, Mammalia
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.