Learn More
PRR5, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PRR5.
Supplier: Abnova Corporation PAB19427
Description
This gene encodes a protein with a proline-rich domain. This gene is located in a region of chromosome 22 reported to contain a tumor suppressor gene that may be involved in breast and colorectal tumorigenesis. Rare read-through transcripts, containing exons from the ARHGAP8 gene which is located immediately downstream, led to the original description of PRR5 and ARHGAP8 as a single gene. Alternative splicing and the use of alternative promoters results in transcripts encoding different isoforms. [provided by RefSeq
Specifications
PRR5 | |
Polyclonal | |
Unconjugated | |
Western Blot (1 ug/mL) Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
PRR5 | |
PRR5 | |
A synthetic peptide corresponding to 20 amino acids near C-terminus of human PRR5. | |
100 μg | |
Primary | |
PRR5 antibody is predicted to not cross-react with other Protor protein family members. | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of PRR5. | |
In PBS (0.02% sodium azide) | |
ARHGAP8/FLJ20185/PP610/PROTOR1 | |
Rabbit | |
Peptide affinity purification | |
RUO | |
55615 | |
Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only