Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRSS1, Rabbit, Clone: A16-N, Abnova™

Catalog No. 89110540 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-540 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-540 Supplier Abnova Corporation Supplier No. MAB1641
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against synthetic peptide of PRSS1 .

This gene encodes a trypsinogen, which is a member of the trypsin family of serine proteases. This enzyme is secreted by the pancreas and cleaved to its active form in the small intestine. It is active on peptide linkages involving the carboxyl group of lysine or arginine. Mutations in this gene are associated with hereditary pancreatitis. This gene and several other trypsinogen genes are localized to the T cell receptor beta locus on chromosome 7. [provided by RefSeq

Specifications

Antigen PRSS1
Applications ELISA, Western Blot
Classification Monoclonal
Clone A16-N
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of PRSS1 .
Dilution Western Blot (1:4000) ELISA (1:20000-1:50000) The optimal working dilution should be determined by the end user.
Formulation In 20mM Tris-HCl, pH 8.0 (10mg/mL BSA, 0.05% sodium azide)
Gene PRSS1
Gene Alias MGC120175/MGC149362/TRP1/TRY1/TRY4/TRYP1
Gene Symbols PRSS1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to N-terminus of human PRSS1 .
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5644
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.