Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTCH1 Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117923 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-923 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-923 Supplier Abnova Corporation Supplier No. PAB15871

Rabbit polyclonal antibody raised against synthetic peptide of Ptch1.

Sequence: KKINYQVDSWE

Specifications

Antigen PTCH1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description patched homolog 1
Formulation In PBS (0.02% sodium azide)
Gene Ptch1
Gene Alias A230106A15Rik, Ptc, Ptc1, Ptch, mes
Gene Symbols Ptch1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 269-279 of mouse Ptch1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19206
Target Species Chicken, Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.