Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTENP1 (phospho S29/T32), Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89121458 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-458 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-458 Supplier Abnova Corporation Supplier No. PAB19954
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant PTENP1.

Specifications

Antigen PTENP1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant PTENP1.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene PTENP1
Gene Alias PTEN-rs/PTEN2/PTH2/psiPTEN
Gene Symbols PTENP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 223-237 of human PTENP1.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11191
Test Specificity Reactivity occurs against human PTENP1(phospho S29/T32) protein and the antibody is specific for the phosphorylated form of the proteins. Approximately 3% reactivity is observed against the non-phosphorylated form of the immunizing peptide. A BLAST analysis was used to suggest cross reactivity with PTEN proteins from mouse, bovine, dog, macaque, chimpanzee and rat sources based on 100% homology with the immunizing sequence.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.