Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTGES/PTGES2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89116771 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-771 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-771 Supplier Abnova Corporation Supplier No. PAB13620
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of PTGES/PTGES2.

Specifications

Antigen PTGES/PTGES2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of PTGES/PTGES2.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In DPBS
Gene Alias MGC10317/MGST-IV/MGST1-L1/MGST1L1/MPGES/PGES/PIG12/PP102/PP1294/TP53I12/mPGES-1
Gene Symbols PTGES
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human PTGES/PTGES2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9536/80142
Test Specificity This antibody is specific to microsomal human PGE Synthase (PGE Isomerase or mPGEi).
Target Species Human
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.