Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTGS1 Rabbit anti-Human, Rat, Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89116772 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-772 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-772 Supplier Abnova Corporation Supplier No. PAB13621
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of PTGS1.

Prostaglandin-endoperoxide synthase (PTGS), also known as cyclooxygenase, is the key enzyme in prostaglandin biosynthesis, and acts both as a dioxygenase and as a peroxidase. There are two isozymes of PTGS: a constitutive PTGS1 and an inducible PTGS2, which differ in their regulation of expression and tissue distribution. This gene encodes PTGS1, which regulates angiogenesis in endothelial cells, and is inhibited by nonsteroidal anti-inflammatory drugs such as aspirin. PTGS1 is thought to be involved in cell-cell signaling and maintaining tissue homeostasis. Alternative splicing of this gene generates two transcript variants. The expression of these two transcripts is differentially regulated by relevant cytokines and growth factors. [provided by RefSeq

Specifications

Antigen PTGS1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of PTGS1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.01% sodium azide
Gene PTGS1
Gene Alias COX1/COX3/PCOX1/PGG/HS/PGHS-1/PGHS1/PHS1/PTGHS
Gene Symbols PTGS1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to PTGS1.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5742
Test Specificity The polyclonal antibody with the highest specificity for Prostaglandin H Synthase 1 (PGHS-1), also known as Cyclooxygenase 1 (COX-1) and corresponds to a sequence region unique to COX-1. This antibody reacts with native, as well as, recombinant COX-1 from several species including human, sheep, rat and most other mammalian species, except mouse. It does not cross-react with COX-2 from any of the species examined to date.
Target Species Human, Rat, Sheep
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.