Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTGS1 Rabbit anti-Bovine, Dog, Human, Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89116773 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-773 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-773 Supplier Abnova Corporation Supplier No. PAB13623
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against PTGS1.

Prostaglandin-endoperoxide synthase (PTGS), also known as cyclooxygenase, is the key enzyme in prostaglandin biosynthesis, and acts both as a dioxygenase and as a peroxidase. There are two isozymes of PTGS: a constitutive PTGS1 and an inducible PTGS2, which differ in their regulation of expression and tissue distribution. This gene encodes PTGS1, which regulates angiogenesis in endothelial cells, and is inhibited by nonsteroidal anti-inflammatory drugs such as aspirin. PTGS1 is thought to be involved in cell-cell signaling and maintaining tissue homeostasis. Alternative splicing of this gene generates two transcript variants. The expression of these two transcripts is differentially regulated by relevant cytokines and growth factors. [provided by RefSeq

Specifications

Antigen PTGS1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against PTGS1.
Dilution ELISA (1:1000) Western Blot (1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene PTGS1
Gene Alias COX1/COX3/PCOX1/PGG/HS/PGHS-1/PGHS1/PHS1/PTGHS
Gene Symbols PTGS1
Host Species Rabbit
Immunogen Sheep PTGS1.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5742
Test Specificity This antibody is reactive in most mammalian species including, human, ovine, bovine, and canine, weakly with mouse and rat. Detects COX-1 expression for human and sheep while reacting weakly with Cyclooxygenase 2 (COX-2).
Target Species Bovine, Canine, Human, Sheep
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.