Learn More
PTGS2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PTGS2.
Supplier: Abnova Corporation PAB12301
Description
Prostaglandin-endoperoxide synthase (PTGS), also known as cyclooxygenase, is the key enzyme in prostaglandin biosynthesis, and acts both as a dioxygenase and as a peroxidase. There are two isozymes of PTGS: a constitutive PTGS1 and an inducible PTGS2, which differ in their regulation of expression and tissue distribution. This gene encodes the inducible isozyme. It is regulated by specific stimulatory events, suggesting that it is responsible for the prostanoid biosynthesis involved in inflammation and mitogenesis. [provided by RefSeq
Specifications
PTGS2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of PTGS2. | |
Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
COX-2/COX2/GRIPGHS/PGG/HS/PGHS-2/PHS-2/hCox-2 | |
Rabbit | |
Affinity purification | |
RUO | |
5743 | |
Human, Mouse, Rat | |
Lyophilized |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Western Blot (1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
PTGS2 | |
PTGS2 | |
A synthetic peptide corresponding to amino acids at N-terminus of human PTGS2. | |
100 μg | |
Primary | |
Different to the related rat sequence by two amino acids. | |
Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only