Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTH2R Rabbit anti-Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114368 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-368 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-368 Supplier Abnova Corporation Supplier No. PAB0720
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Pth2r.

Sequence: RQIDSHVTLPGYVWSSSEQDC

Specifications

Antigen PTH2R
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Pth2r.
Dilution Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene Pth2r
Gene Alias Pthr2
Gene Symbols Pth2r
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 481-501 of rat Pth2r.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81753
Target Species Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.