Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTK2 Rabbit anti-Bovine, Chicken, Dog, Hamster, Human, Monkey, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89118175 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-175 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-175 Supplier Abnova Corporation Supplier No. PAB16301
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of PTK2.

This gene encodes a cytoplasmic protein tyrosine kinase which is found concentrated in the focal adhesions that form between cells growing in the presence of extracellular matrix constituents. The encoded protein is a member of the FAK subfamily of protein tyrosine kinases but lacks significant sequence similarity to kinases from other subfamilies. Activation of this gene may be an important early step in cell growth and intracellular signal transduction pathways triggered in response to certain neural peptides or to cell interactions with the extracellular matrix. At least four transcript variants encoding four different isoforms have been found for this gene, but the full-length natures of only two of them have been determined. [provided by RefSeq

Specifications

Antigen PTK2
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of PTK2.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (13 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene PTK2
Gene Alias FADK/FAK/FAK1/pp125FAK
Gene Symbols PTK2
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to human PTK2.
Purification Method Immunoaffinity purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5747
Test Specificity Kinase domain of human.
Target Species Bovine, Canine, Chicken, Hamster, Human, Monkey, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.