Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTP4A2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120455 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-455 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-455 Supplier Abnova Corporation Supplier No. PAB18892
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant PTP4A2.

The protein encoded by this gene belongs to a small class of the protein tyrosine phosphatase (PTP) family. PTPs are cell signaling molecules that play regulatory roles in a variety of cellular processes. PTPs in this class contain a protein tyrosine phosphatase catalytic domain and a characteristic C-terminal prenylation motif. This PTP has been shown to primarily associate with plasmic and endosomal membrane through its C-terminal prenylation. This PTP was found to interact with the beta-subunit of Rab geranylgeranyltransferase II (beta GGT II), and thus may function as a regulator of GGT II activity. Overexpression of this gene in mammalian cells conferred a transformed phenotype, which suggested its role in tumorigenesis. Alternatively spliced transcript variants that encode two distinct isoforms have been described. [provided by RefSeq

Specifications

Antigen PTP4A2
Applications ELISA, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant PTP4A2.
Dilution ELISA (1:10000-1:80000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide
Gene PTP4A2
Gene Alias HH13/HH7-2/HU-PP-1/OV-1/PRL-2/PRL2/PTP4A/PTPCAAX2/ptp-IV1a/ptp-IV1b
Gene Symbols PTP4A2
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 2-167 of human PTP4A2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8073
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.