Learn More
PTPN6 (phospho Y536), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of PTPN6.
Supplier: Abnova Corporation PAB7923
Description
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. N-terminal part of this PTP contains two tandem Src homolog (SH2) domains, which act as protein phospho-tyrosine binding domains, and mediate the interaction of this PTP with its substrates. This PTP is expressed primarily in hematopoietic cells, and functions as an important regulator of multiple signaling pathways in hematopoietic cells. This PTP has been shown to interact with, and dephosphorylate a wide spectrum of phospho-proteins involved in hematopoietic cell signaling. Multiple alternatively spliced variants of this gene, which encode distinct isoforms, have been reported. [provided by RefSeq
Specifications
PTPN6 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of PTPN6. | |
In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide) | |
HCP/HCPH/HPTP1C/PTP-1C/SH-PTP1/SHP-1/SHP-1L/SHP1 | |
Rabbit | |
100 μL | |
Primary | |
The sequence is highly conserved in rat and mouse SHP1. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:2000) Western Blot (1:500) The optimal working dilution should be determined by the end user. | |
PTPN6 | |
PTPN6 | |
Synthetic phosphopeptide corresponding to residues surrounding Y536 of human PTPN6. | |
RUO | |
5777 | |
Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only