Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPRZ1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89122295 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-122-295 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-122-295 Supplier Abnova Corporation Supplier No. PAB20795
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant PTPRZ1.

This gene is a member of the receptor type protein tyrosine phosphatase family and encodes a single-pass type I membrane protein with two cytoplasmic tyrosine-protein phosphatase domains, an alpha-carbonic anhydrase domain and a fibronectin type-III domain. Expression of this gene is induced in gastric cancer cells, in the remyelinating oligodendrocytes of multiple sclerosis lesions, and in human embryonic kidney cells under hypoxic conditions. Both the protein and transcript are overexpressed in glioblastoma cells, promoting their haptotactic migration. Alternative splice variants that encode different protein isoforms have been described but their full-length nature has not been determined. [provided by RefSeq

Sequence: LFRHLHTVSQILPQVTSATESDKVPLHASLPVAGGDLLLEPSLAQYSDVLSTTHAASETLEFGSESGVLYKTLMFSQVEPPSSDAMMHARSSGPEPSYALSDNEGSQHIFTVSYSSAIPVHDSVGVTYQGSLFSGPSHIPIPKSSLIT

Specifications

Antigen PTPRZ1
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant PTPRZ1.
Dilution Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene PTPRZ1
Gene Alias HPTPZ/HPTPzeta/PTP-ZETA/PTP18/PTPRZ/PTPZ/RPTPB/RPTPbeta/phosphacan
Gene Symbols PTPRZ1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human PTPRZ1.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5803
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.