Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTTG1/PTTG2/PTTG3P, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89126550 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-126-550 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-126-550 Supplier Abnova Corporation Supplier No. PAB24878
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of PTTG1/PTTG2/PTTG3P.

Specifications

Antigen PTTG1/PTTG2/PTTG3P
Applications Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of PTTG1/PTTG2/PTTG3P.
Dilution Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In 1X PBS, pH 7.2 (0.05% sodium azide)
Gene Alias EAP1/HPTTG/MGC126883/MGC138276/PTTG/TUTR1
Gene Symbols PTTG1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to PTTG1/PTTG2/PTTG3P.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9232/10744/26255
Test Specificity PTTG1/PTTG2/PTTG3P polyclonal antibody detects endogenous levels of PTTG1/PTTG2/PTTG3P protein.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.