Learn More
PXK, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PXK.
Supplier: Abnova Corporation PAB4535
Description
PXK binds to the Na,K-ATPase beta-1 (ATP1B1; MIM 182330) and beta-3 (ATP1B3; MIM 601867) subunits and modulates both Na,K-ATPase enzymatic and ion pump activities (Mao et al., 2005 [PubMed 16135750]).[supplied by OMIM
Specifications
PXK | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of PXK. | |
In PBS (0.09% sodium azide) | |
FLJ20335/MONaKA | |
Rabbit | |
Protein G purification | |
RUO | |
54899 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:1000) Western Blot (1:100-500) The optimal working dilution should be determined by the end user. | |
PXK | |
PXK | |
A synthetic peptide (conjugated with KLH) corresponding to internal region of human PXK. | |
400 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only