Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyruvate Dehydrogenase E1-alpha subunit, p Ser293 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11093479AF700
Description
Pyruvate Dehydrogenase E1-alpha subunit Polyclonal specifically detects Pyruvate Dehydrogenase E1-alpha subunit in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoblotting, Knockout Validated.Specifications
Pyruvate Dehydrogenase E1-alpha subunit (p Ser293) | |
Polyclonal | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoblotting, Knockout Validated | |
P08559 | |
PDHA1 | |
A synthetic peptide surrounding the phosphorylated serine 293 of the human Pyruvate Dehydrogenase E1-alpha subunit protein. [Swiss-Prot: #P08559] | |
Affinity Purified | |
RUO | |
Primary | |
This is specific for the phosphorylated Serine 293 form of the PDHE1 alpha protein. | |
Store at 4C in the dark. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoblot, Immunoassay | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EC 1.2.4.1, PDHA, PDHE1-A type I, PHE1APDHCE1A, pyruvate dehydrogenase (lipoamide) alpha 1, pyruvate dehydrogenase complex, E1-alpha polypeptide 1, pyruvate dehydrogenase E1 component subunit alpha, somatic form, mitochondrial | |
Rabbit | |
43 kDa | |
0.05 mL | |
Cancer, Core ESC Like Genes, Lipid and Metabolism, Phospho Specific, Signal Transduction, Stem Cell Markers | |
5160 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction